N-(6-Aminohexyl)-1-naphthalenesulphonamide hydrochloride ≥98.0% (通过HPLC,总氮)

供应商: TCI

A2410-25MG A2410-500MG
TCIAA2410-25MGEA 0 CNY
TCIAA2410-25MG TCIAA2410-500MG
N-(6-Aminohexyl)-1-naphthalenesulphonamide hydrochloride ≥98.0% (通过HPLC,总氮)
N-(6-Aminohexyl)-1-naphthalenesulphonamide hydrochloride
分子式: C₁₆H₂₂N₂O₂S
存储条件: Freezer
MDL编号: MFCD00058043
CAS号: 61714-25-8

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 162.0 to 166.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR