3-氨基-5-甲基吡啶 ≥97.0% (by GC, titration analysis)

供应商: TCI
Danger

Synonyms: 5-Amino-3-picoline

A2396-1G A2396-5G
TCIAA2396-1GEA 0 CNY
TCIAA2396-1G TCIAA2396-5G
3-氨基-5-甲基吡啶 ≥97.0% (by GC, titration analysis)
3-氨基-5-甲基吡啶
分子式: C6H8N2
分子量: 108.1411 g/mol
沸点: 155°C
MDL编号: MFCD04112508
CAS号: 3430-19-1
UN号: 2811
ADR: 6.1 III

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 63.0 to 67.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR