Aniracetam ≥98.0% (by GC)

供应商: TCI

Synonyms: 1-(4-Methoxybenzoyl)pyrrolidin-2-one, 1-(4-Methoxybenzoyl)-2-pyrrolidone

A2394-5G A2394-1G
TCIAA2394-5GEA 0 CNY
TCIAA2394-5G TCIAA2394-1G
Aniracetam ≥98.0% (by GC)
Aniracetam
分子式: C₁₂H₁₃NO₃
MDL编号: MFCD00153767
CAS号: 72432-10-1
Merck Index: 14,00662

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 120.0 to 124.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR