(S)-2-Acetamido-3-methylbutanoic acid ≥98.0% (by HPLC, titration analysis)

供应商: TCI

Synonyms: N-Ac-Val-OH, Ac-Val-OH, N-Acetyl-L-Valine

A2366-25G A2366-5G
TCIAA2366-25GEA 0 CNY
TCIAA2366-25G TCIAA2366-5G
(S)-2-Acetamido-3-methylbutanoic acid ≥98.0% (by HPLC, titration analysis)
(S)-2-Acetamido-3-methylbutanoic acid
分子式: C₇H₁₃NO₃
MDL编号: MFCD00066066
CAS号: 96-81-1
EINECS: 202-537-8

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 163.0 to 167.0 deg-C
Specific rotation [a]20/D +11.0 to +14.0 deg (C=1, AcOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR