O-Acetyl-N-methylhydroxylamine hydrochloride ≥98.0% (by total nitrogen and titration analysis)

供应商: TCI

A2365-1G A2365-5G
TCIAA2365-1GEA 0 CNY
TCIAA2365-1G TCIAA2365-5G
O-Acetyl-N-methylhydroxylamine hydrochloride ≥98.0% (by total nitrogen and titration analysis)
O-Acetyl-N-methylhydroxylamine hydrochloride
分子式: C₃H₇NO₂
分子量: 125.55 g/mol
溶点: 111°C
存储条件: Refrigerator
CAS号: 19689-95-3

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR