9-蒽硼酸 ≥98.0% (通过HPLC) (contains varying amounts of Anhydride)
供应商: TCI
|
Warning
|
A2328-1G
A2328-5G
TCIAA2328-1GEA
0
CNY
TCIAA2328-1G
TCIAA2328-5G
9-蒽硼酸 ≥98.0% (通过HPLC) (contains varying amounts of Anhydride)
9-蒽硼酸
分子式:
C₁₄H₁₁BO₂ 溶点: 217°C |
MDL编号:
MFCD03425925 CAS号: 100622-34-2 |
产品技术规格
Appearance | Light yellow to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...