4-Azidosalicylic acid ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Warning

Synonyms: 4-Azido-2-hydroxybenzoic Acid

A2290-1G A2290-100MG
TCIAA2290-1GEA 0 CNY
TCIAA2290-1G TCIAA2290-100MG
4-Azidosalicylic acid ≥98.0% (by HPLC, titration analysis)
4-Azidosalicylic acid
分子式: C₇H₅N₃O₃
分子量: 179.14 g/mol
溶点: 197°C
存储条件: Refrigerator
MDL编号: MFCD07367665
CAS号: 66761-27-1
UN号: 1325
ADR: 4.1 III

现在订购

产品技术规格

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR