4-(2-Aminoethyl)cyclohexanol cis- and trans-mixture ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Danger
|
Synonyms:
4-(2-Aminoethyl)cyclohexanol (cis- and trans- mixture)
A2266-5G
A2266-1G
TCIAA2266-5GEA
0
CNY
TCIAA2266-5G
TCIAA2266-1G
4-(2-Aminoethyl)cyclohexanol cis- and trans-mixture ≥98.0% (by GC, titration analysis)
4-(2-Aminoethyl)cyclohexanol
分子式:
C₈H₁₇NO 分子量: 143.23 g/mol 沸点: 256°C |
MDL编号:
MFCD11520548 CAS号: 148356-06-3 UN号: 2735 ADR: 8 III |
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % (total of isomers) |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...