Alendronic acid ≥98.0% (通过滴定分析)

供应商: TCI
Danger

A2120-25G A2120-5G
TCIAA2120-25GEA 0 CNY
TCIAA2120-25G TCIAA2120-5G
Alendronic acid ≥98.0% (通过滴定分析)
Alendronic acid
分子式: C₄H₁₃NO₇P₂
溶点: 235°C
MDL编号: MFCD00868112
CAS号: 66376-36-1
UN号: 1759
ADR: 8,III
Merck Index: 14,00229

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR