(S)-4-(4-氨基苄基)-1,3-唑烷-2-酮 ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: (4S)-4-[(4-氨基苯基)甲基]-2-恶唑烷酮

A2095-1G A2095-5G
TCIAA2095-1GEA 0 CNY
TCIAA2095-1G TCIAA2095-5G
(S)-4-(4-氨基苄基)-1,3-唑烷-2-酮 ≥98.0% (by GC, titration analysis)
(S)-4-(4-氨基苄基)-1,3-唑烷-2-酮
分子式: C₁₀H₁₂N₂O₂
分子量: 192.22 g/mol
MDL编号: MFCD03411476
CAS号: 152305-23-2

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 108.0 to 112.0 deg-C
Specific rotation [a]20/D -4.0 to -7.0 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR