3-Amino-1-adamantanol hydrate ≥98.0% (通过GC,滴定分析)

供应商: TCI
Warning

A2079-1G A2079-5G
TCIAA2079-1GEA 0 CNY
TCIAA2079-1G TCIAA2079-5G
3-Amino-1-adamantanol hydrate ≥98.0% (通过GC,滴定分析)
3-Amino-1-adamantanol hydrate
分子式: C₁₀H₁₇NO·xH₂O
溶点: 269°C
MDL编号: MFCD01821204
CAS号: 702-82-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6da9a0769b06e696c708e4e39c7a9659ddbb30f&vid=5b682a85fa5f6684630c0404f2a9999dedd0214a&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Water max. 4.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR