7-Aminodesacetoxycephalosporanic acid ≥98.0% (通过HPLC)

供应商: TCI
Danger

A2075-25G A2075-5G
TCIAA2075-25GEA 0 CNY
TCIAA2075-25G TCIAA2075-5G
7-Aminodesacetoxycephalosporanic acid ≥98.0% (通过HPLC)
7-Aminodesacetoxycephalosporanic acid
分子式: C₈H₁₀N₂O₃S
溶点: 242°C
MDL编号: MFCD00151456
CAS号: 22252-43-3
EINECS: 244-870-1

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystalline
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 95.0 %
Specific rotation [a]20/D +100.0 to +107.0 deg (C=1, 1mol/L HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR