1H-Pyrazole-1-carboximidamide hydrochloride ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Danger

Synonyms: 1-Amidinopyrazole Hydrochloride, 1H-Pyrazole-1-carboxamidine monohydrochloride, Hydrochloride, 1-Pyrazolecarboxamidine, Praxadine

A2055-25G A2055-5G
TCIAA2055-25GEA 0 CNY
TCIAA2055-25G TCIAA2055-5G
1H-Pyrazole-1-carboximidamide hydrochloride ≥98.0% (by HPLC, titration analysis)
1H-Pyrazole-1-carboximidamide hydrochloride
分子式: C₄H₇ClN₄
分子量: 146.58 g/mol
MDL编号: MFCD00210087
CAS号: 4023-02-3
EINECS: 429-520-1

现在订购

产品技术规格

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 165.0 to 169.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR