9-Acetyl-3,6-diiodocarbazole ≥96.0% (by total nitrogen basis)

供应商: TCI

Synonyms: 1-(3,6-Diiodo-9H-carbazol-9-yl)ethanone

A2051-1G A2051-5G
TCIAA2051-1GEA 0 CNY
TCIAA2051-1G TCIAA2051-5G
9-Acetyl-3,6-diiodocarbazole ≥96.0% (by total nitrogen basis)
9-Acetyl-3,6-diiodocarbazole
分子式: C₁₄H₉I₂NO
MDL编号: MFCD10000947
CAS号: 606129-89-9

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystalline
Purity (with Total Nitrogen) min. 96.0 %
Melting point 225.0 to 231.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR