(R)-tert-Butyl-2-aminopropanoate hydrochloride ≥98.0% (通过HPLC,总氮)

供应商: TCI

A2009-1G A2009-5G
TCIAA2009-1GEA 0 CNY
TCIAA2009-1G TCIAA2009-5G
(R)-tert-Butyl-2-aminopropanoate hydrochloride ≥98.0% (通过HPLC,总氮)
(R)-tert-Butyl-2-aminopropanoate hydrochloride
分子式: C₇H₁₅NO₂·HCl
MDL编号: MFCD00077087
CAS号: 59531-86-1

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) (CAD) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Specific rotation [a]20/D -1.0 to -2.0 deg (C=2, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR