6-Amino-2,4-dichloro-3-methylphenol hydrochloride ≥98.0% (by HPLC)
供应商: TCI
|
Warning
|
Synonyms:
6-Amino-2,4-dichloro-m-cresol Hydrochloride
A1998-25G
A1998-5G
TCIAA1998-25GEA
0
CNY
TCIAA1998-25G
TCIAA1998-5G
6-Amino-2,4-dichloro-3-methylphenol hydrochloride ≥98.0% (by HPLC)
6-Amino-2,4-dichloro-3-methylphenol hydrochloride
分子式:
C₇H₇Cl₂NO |
MDL编号:
MFCD00035472 CAS号: 39549-31-0 EINECS: 254-503-7 |
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...