5-Amino-2-(4-aminophenyl)benzofuran ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 2-(4-Aminophenyl)benzofuran-5-amine, 2-(4-Aminophenyl)-5-benzofuranamine

A1966-1G A1966-5G
TCIAA1966-1GEA 0 CNY
TCIAA1966-1G TCIAA1966-5G
5-Amino-2-(4-aminophenyl)benzofuran ≥98.0% (by GC)
5-Amino-2-(4-aminophenyl)benzofuran
分子式: C₁₄H₁₂N₂O
分子量: 224.26 g/mol
MDL编号: MFCD10000946
CAS号: 84102-58-9

现在订购

产品技术规格

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 197.0 to 201.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR