阿苯达唑 ≥98.0% (by total nitrogen basis)

供应商: TCI
Danger

Synonyms: 5-(丙硫基)-2-苯并咪唑氨基甲酸甲酯

A1943-25G A1943-5G
TCIAA1943-25GEA 0 CNY
TCIAA1943-25G TCIAA1943-5G
阿苯达唑 ≥98.0% (by total nitrogen basis)
阿苯达唑
分子式: C₁₂H₁₅N₃O₂S
分子量: 265.33 g/mol
溶点: 210°C
MDL编号: MFCD00083232
CAS号: 54965-21-8
EINECS: 259-414-7
Merck Index: 14,00210

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR