Acyclovir ≥98.0% (by HPLC)
供应商: TCI
|
Warning
|
Synonyms:
1,9-dihydro-2-amino-9-((2-hydroxyethoxy)methyl)-6h-purin-6-on, Acycloguanosine, 2-amino-1,9-dihydro-9-((2-hydroxyethoxy)methyl)-6h-purin-6-one, Acyclovir, 9-[(2-Hydroxyethoxy)methyl]guanine, 2-Amino-1,9-dihydro-9-((2-hydroxyethoxy)methyl)-6H-purin-6-one, 2-Amino-1,9-dihydro-9-[(2-hydroxyethoxy)methyl]-6H-purin-6-one, Aciclovir, 2-Amino-9-((2-hydroxyethoxy)methyl)-1H-purin-6(9H)-one
A1915-25G
A1915-5G
TCIAA1915-25GEA
0
CNY
TCIAA1915-25G
TCIAA1915-5G
Acyclovir ≥98.0% (by HPLC)
Acyclovir
分子式:
C₈H₁₁N₅O₃ 分子量: 225.21 g/mol 溶点: 262°C |
MDL编号:
MFCD00057880 CAS号: 59277-89-3 EINECS: 261-685-1 Merck Index: 14,00146 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 94.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...