1-Aminohydantoin hydrochloride ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 1-Aminohydantoin Hydrochloride, 1-Aminohydantoin HCl, 1-Amino-2,4-imidazolidinedione hydrochloride

A1902-25G A1902-5G
TCIAA1902-25GEA 0 CNY
TCIAA1902-25G TCIAA1902-5G
1-Aminohydantoin hydrochloride ≥98.0% (by titrimetric analysis)
1-Aminohydantoin hydrochloride
分子式: C₃H₅N₃O₂
分子量: 151.55 g/mol
溶点: 203°C
MDL编号: MFCD02181047
CAS号: 2827-56-7

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR