N-Aminosuccinimide hydrochloride ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

A1819-1G A1819-5G
TCIAA1819-1GEA 0 CNY
TCIAA1819-1G TCIAA1819-5G
N-Aminosuccinimide hydrochloride ≥98.0% (by titrimetric analysis)
N-Aminosuccinimide hydrochloride
分子式: C₄H₆N₂O₂
分子量: 150.56 g/mol
存储条件: Refrigerator
CAS号: 108906-15-6

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR