2-Acetamido-4,6-O-benzylidene-2-deoxy-β-D-glucopyranosyl azide ≥98.0% (by HPLC)

供应商: TCI

Synonyms: 2-​Acetamido-​4,​6-​O-​benzylidene-​2-​deoxy-​β-​D-​glucopyranosyl azide

A1811-1G A1811-5G
TCIAA1811-1GEA 0 CNY
TCIAA1811-1G TCIAA1811-5G
2-Acetamido-4,6-O-benzylidene-2-deoxy-β-D-glucopyranosyl azide ≥98.0% (by HPLC)
2-Acetamido-4,6-O-benzylidene-2-deoxy-beta-D-glucopyranosyl azide
分子式: C₁₅H₁₈N₄O₅
分子量: 334.33 g/mol
存储条件: Freezer
CAS号: 168397-51-1

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D -95.0 to -99.0 deg (C=0.5, Methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR