Azobenzene-3,3'-dicarboxylic acid ≥95.0% (by HPLC, titration analysis)

供应商: TCI
Warning

Synonyms: Azobenzene-3,3'-dicarboxylic Acid, 3,3'-Azodibenzoic Acid, 3,3'-Azodibenzoic acid, 3,3'-(Diazene-1,2-diyl)dibenzoic acid

A1598-1G A1598-5G
TCIAA1598-1GEA 0 CNY
TCIAA1598-1G TCIAA1598-5G
Azobenzene-3,3'-dicarboxylic acid ≥95.0% (by HPLC, titration analysis)
Azobenzene-3,3'-dicarboxylic acid
分子式: C₁₄H₁₀N₂O₄
分子量: 270.24 g/mol
MDL编号: MFCD03844759
CAS号: 621-18-1

现在订购

产品技术规格

Appearance Light yellow to Amber to Dark green powder to crystaline
Purity (HPLC) min. 95.0 area%
Purity (Neutralization titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR