N-甲基-4-氨基邻苯二甲酰亚胺 ≥98.0% (by HPLC, total nitrogen)

供应商: TCI
Warning

Synonyms: 5-Amino-2-methylisoindoline-1,3-dione

A1561-25G A1561-5G
TCIAA1561-25GEA 0 CNY
TCIAA1561-25G TCIAA1561-5G
N-甲基-4-氨基邻苯二甲酰亚胺 ≥98.0% (by HPLC, total nitrogen)
N-甲基-4-氨基邻苯二甲酰亚胺
分子式: C₉H₈N₂O₂
分子量: 176.18 g/mol
MDL编号: MFCD00012081
CAS号: 2307-00-8

现在订购

产品技术规格

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 244.0 to 248.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR