4-(2-Aminoethyl)-1-benzylpiperidine ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 2-(1-Benzyl-4-piperidyl)ethylamine

A1517-25G A1517-5G
TCIAA1517-25GEA 0 CNY
TCIAA1517-25G TCIAA1517-5G
4-(2-Aminoethyl)-1-benzylpiperidine ≥98.0% (by GC, titration analysis)
4-(2-Aminoethyl)-1-benzylpiperidine
分子式: C₁₄H₂₂N₂
分子量: 218.34 g/mol
MDL编号: MFCD02093423
CAS号: 86945-25-7

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR