(S)-Methyl-2-aminopropanoate hydrochloride ≥98.0% (by titrimetric analysis)

供应商: TCI

Synonyms: H-Ala-OMe·HCl, H-Ala-OMe.HCl, L-Alanine methyl ester hydrochloride, (S)-2-Aminopropionic Acid Methyl Ester Hydrochloride

A1466-25G A1466-5G
TCIAA1466-25GEA 0 CNY
TCIAA1466-25G TCIAA1466-5G
(S)-Methyl-2-aminopropanoate hydrochloride ≥98.0% (by titrimetric analysis)
(S)-Methyl-2-aminopropanoate hydrochloride
分子式: C₄H₉NO₂·HCl
分子量: 139.6 g/mol
MDL编号: MFCD00063663
CAS号: 2491-20-5
EINECS: 219-652-4

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Melting point 108.0 to 112.0 deg-C
Specific rotation [a]20/D +5.0 to +8.0 deg (C=2, MeOH)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR