O-Allylhydroxylamine hydrochloride ≥98.0% (通过滴定分析)

供应商: TCI
Warning

A1449-1G A1449-5G
TCIAA1449-1GEA 0 CNY
TCIAA1449-1G TCIAA1449-5G
O-Allylhydroxylamine hydrochloride ≥98.0% (通过滴定分析)
O-Allylhydroxylamine hydrochloride
分子式: C₃H₇NO
MDL编号: MFCD00012957
CAS号: 38945-21-0
EINECS: 254-203-6

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR