β-Alanine benzyl ester p-toluenesulphonate ≥98.0% (by HPLC, titration analysis)

供应商: TCI

Synonyms: H-β-Ala-OBzl·p-tosylate, H-β-Ala-Obzl.TosOH, H-β-Ala-OBzl·TosOH

A1423-25G A1423-5G
TCIAA1423-25GEA 0 CNY
TCIAA1423-25G TCIAA1423-5G
β-Alanine benzyl ester p-toluenesulphonate ≥98.0% (by HPLC, titration analysis)
β-Alanine benzyl ester p-toluenesulphonate
分子式: C₁₀H₁₃NO₂
分子量: 351.42 g/mol
MDL编号: MFCD00039061
CAS号: 27019-47-2

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 % (total of Ion-Pair)
Purity (Neutralization titration) min. 98.0 %
Solubility in hot Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR