Acetyl-L-carnitine hydrochloride ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: (R)-3-Acetoxy-4-(trimethylammonio)butyrate Hydrochloride, (R)-2-Acetoxy-3-carboxy-N,N,N-trimethylpropan-1-aminium chloride, Acetyl-L-carnitine HCl

A1394-25G A1394-5G
TCIAA1394-25GEA 0 CNY
TCIAA1394-25G TCIAA1394-5G
Acetyl-L-carnitine hydrochloride ≥98.0% (by titrimetric analysis)
Acetyl-L-carnitine hydrochloride
分子式: C₉H₁₇NO₄
分子量: 239.70 g/mol
MDL编号: MFCD00082230
CAS号: 5080-50-2
Merck Index: 14,00084

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Specific rotation [a]20/D -27.0 to -29.0 deg (C=1, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR