Heptylnonyl adipate (so called) plasticizer

供应商: TCI

Synonyms: Adipic Acid Heptylnonyl Ester

A1388-25ML A1388-500ML
TCIAA1388-25MLEA 0 CNY
TCIAA1388-25ML TCIAA1388-500ML
Heptylnonyl adipate (so called) plasticizer
Heptylnonyl adipate
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=512b852664a38e4a4a39136ae8145c0a5b9bdd5e&vid=dc4454559d5359f49933baf2d7d6888d5b6dffcf&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) mixture of Heptylnonyl adipate, Diheptyl adipate, Dinonyl adipate


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR