Diisononyl adipate plasticizer

供应商: TCI

Synonyms: Adipic Acid Diisononyl Ester

A1387-25ML A1387-500ML
TCIAA1387-25MLEA 0 CNY
TCIAA1387-25ML TCIAA1387-500ML
Diisononyl adipate plasticizer
Diisononyl adipate
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a35bfb9ca3aa7ff0f57437ce0497214b451ba8b&vid=3f3729ae34a32f229539594b8344e46dcfb20543&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) many component mixture


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR