1,3-金刚烷二甲酸 ≥97.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: ADC

A1358-25G A1358-5G
TCIAA1358-25GEA 0 CNY
TCIAA1358-25G TCIAA1358-5G
1,3-金刚烷二甲酸 ≥97.0% (by GC, titration analysis)
1,3-金刚烷二甲酸
分子式: C₁₂H₁₆O₄
分子量: 224.26 g/mol
MDL编号: MFCD00167790
CAS号: 39269-10-8
EINECS: 254-395-1

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Solubility in hot Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR