2-Propoxyethylamine ≥97.0% (by GC, titration analysis)

供应商: TCI
Danger

Synonyms: 2-Aminoethylpropylether, 2-Aminoethyl Propyl Ether, O-Propylethanolamine

A1325-25ML A1325-5ML
TCIAA1325-25MLEA 0 CNY
TCIAA1325-25ML TCIAA1325-5ML
2-Propoxyethylamine ≥97.0% (by GC, titration analysis)
2-Propoxyethylamine
分子式: C₅H₁₃NO
分子量: 103.17 g/mol
沸点: 126°C
MDL编号: MFCD00047955
CAS号: 42185-03-5
EINECS: 255-699-7
UN号: 2733
ADR: 3 III

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR