壬二酸氢甲酯 ≥97.0% (by GC, titration analysis)

供应商: TCI

Synonyms: mono-Methyl nonanedioate, 壬二酸单甲酯, Azelaic acid monomethyl ester

A1312-25G A1312-5G
TCIAA1312-25GEA 0 CNY
TCIAA1312-25G TCIAA1312-5G
壬二酸氢甲酯 ≥97.0% (by GC, titration analysis)
壬二酸氢甲酯
分子式: C₁₀H₁₈O₄
分子量: 202.25 g/mol
沸点: 160°C
存储条件: Freezer
MDL编号: MFCD00004431
CAS号: 2104-19-0
EINECS: 218-275-2

现在订购

产品技术规格

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) 97.0 to 103.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR