2-乙酰胺基丙烯酸甲酯 ≥97.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
2-Acetamidoacrylic Acid Methyl Ester, Methyl 2-acetamidoacrylate
A1304-1G
A1304-5G
TCIAA1304-1GEA
0
CNY
TCIAA1304-1G
TCIAA1304-5G
2-乙酰胺基丙烯酸甲酯 ≥97.0% (by GC)
2-乙酰胺基丙烯酸甲酯
分子式:
C₆H₉NO₃ 分子量: 143.14 g/mol 沸点: 104°C 存储条件: Refrigerator |
MDL编号:
MFCD00013394 CAS号: 35356-70-8 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 50.0 to 53.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...