2-Amino-3-chloro-1,4-naphthoquinone ≥98.0% (by HPLC, total nitrogen)

供应商: TCI
Warning

A1288-25G A1288-500G
TCIAA1288-25GEA 0 CNY
TCIAA1288-25G TCIAA1288-500G
2-Amino-3-chloro-1,4-naphthoquinone ≥98.0% (by HPLC, total nitrogen)
2-Amino-3-chloro-1,4-naphthoquinone
分子式: C₁₀H₆ClNO₂
分子量: 207.61 g/mol
MDL编号: MFCD00001680
CAS号: 2797-51-5
EINECS: 220-529-2

现在订购

产品技术规格

Appearance Orange to Amber to Dark red powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 199.0 to 203.0 deg-C
Solubility in N,N-DMF almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR