Tris(trifluoro-2,4-pentanedionato)aluminum(III) ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: Aluminum(III) Trifluoroacetylacetonate, Trifluoroacetylacetonato Aluminum(III) Salt

A1267-25G A1267-5G
TCIAA1267-25GEA 0 CNY
TCIAA1267-25G TCIAA1267-5G
Tris(trifluoro-2,4-pentanedionato)aluminum(III) ≥98.0% (by titrimetric analysis)
Tris(trifluoro-2,4-pentanedionato)aluminum(III)
分子式: C₁₅H₁₂AlF₉O₆
分子量: 486.22 g/mol
MDL编号: MFCD00042062
CAS号: 14354-59-7

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (Chelometric Titration) min. 98.0 %
Melting point 119.0 to 122.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR