Dimethyl-4-acetoxyisophthalate ≥98.0% (by GC)

供应商: TCI

Synonyms: 4-Acetoxyisophthalic Acid Dimethyl Ester

A1226-1G A1226-5G
TCIAA1226-1GEA 0 CNY
TCIAA1226-1G TCIAA1226-5G
Dimethyl-4-acetoxyisophthalate ≥98.0% (by GC)
Dimethyl-4-acetoxyisophthalate
分子式: C₁₂H₁₂O₆
MDL编号: MFCD00143241
CAS号: 71932-29-1

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 93.0 to 98.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR