1,6-脱水-β-D-葡萄糖 ≥99.0% (by GC)
供应商: TCI
Synonyms:
左旋葡聚糖
A1074-1G
A1074-5G
TCIAA1074-1GEA
0
CNY
TCIAA1074-1G
TCIAA1074-5G
1,6-脱水-β-D-葡萄糖 ≥99.0% (by GC)
1,6-脱水-β-D-葡萄糖
分子式:
C₆H₁₀O₅ |
MDL编号:
MFCD00063248 CAS号: 498-07-7 EINECS: 207-855-0 |
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 177.0 to 184.0 deg-C |
Specific rotation [a]20/D | -65.0 to -68.0 deg (C=2, H2O) |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...