Allura red AC

供应商: TCI

A0943-25G A0943-5G
TCIAA0943-25GEA 0 CNY
TCIAA0943-25G TCIAA0943-5G
Allura red AC
Allura red AC
分子式: C₁₈H₁₄N₂O₈S₂·2Na
MDL编号: MFCD00059526
CAS号: 25956-17-6
EINECS: 247-368-0
Merck Index: 14,00284

现在订购

产品技术规格

Appearance Orange to Brown to Dark red powder to crystal
Lambda max. 497.0 to 501.0 nm (0.02mol/L AcONH4 sol.)
Absorbance (E1%1cm) min. 400 (0.02mol/L AcONH4 sol., 497.0 to 501.0 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR