N-(3-氨丙基)-2-甲基哌啶 ≥98.0% (by GC, titration analysis)

供应商: TCI
Danger

Synonyms: 1-(3-Aminopropyl)-2-methylhexahydropyridine, 2-Methyl-1-piperidinepropanamine, N-(3-Aminopropyl)-2-pipecoline, 3-(2-Methylpiperidin-1-yl)propan-1-amine, 1-(3-Aminopropyl)-2-methylpiperidine, 1-(3-Aminopropyl)-2-pipecoline

A0833-25ML A0833-500ML
TCIAA0833-25MLEA 0 CNY
TCIAA0833-25ML TCIAA0833-500ML
N-(3-氨丙基)-2-甲基哌啶 ≥98.0% (by GC, titration analysis)
N-(3-氨丙基)-2-甲基哌啶
分子式: C₉H₂₀N₂
分子量: 156.27 g/mol
沸点: 170°C
闪点: 88°C
MDL编号: MFCD00006517
CAS号: 25560-00-3
EINECS: 247-102-3
UN号: 3267
ADR: 8 III

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR