Ethyl-1-adamantanecarboxylate ≥98.0% (by GC)

供应商: TCI

Synonyms: 1-Adamantanecarboxylic Acid Ethyl Ester, Ethyl adamantane-1-carboxylate

A0722-25G A0722-5G
TCIAA0722-25GEA 0 CNY
TCIAA0722-25G TCIAA0722-5G
Ethyl-1-adamantanecarboxylate ≥98.0% (by GC)
Ethyl-1-adamantanecarboxylate
分子式: C₁₃H₂₀O₂
分子量: 208.30 g/mol
沸点: 123°C
MDL编号: MFCD00078272
CAS号: 2094-73-7
EINECS: 218-253-2

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR