去乙基-去异丙基-2-羟基阿特拉津 ≥95.0% (by total nitrogen basis)
供应商: TCI
Synonyms:
4,6-二氨基-2-羟基-1,3,5-三嗪, 三聚氰胺二酰胺
A0676-25G
A0676-5G
TCIAA0676-25GEA
0
CNY
TCIAA0676-25G
TCIAA0676-5G
去乙基-去异丙基-2-羟基阿特拉津 ≥95.0% (by total nitrogen basis)
去乙基-去异丙基-2-羟基阿特拉津
分子式:
C₃H₅N₅O 分子量: 127.11 g/mol |
MDL编号:
MFCD00023186 CAS号: 645-92-1 EINECS: 211-455-1 |
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (with Total Nitrogen) | min. 95.0 % |
Solubility in hot 1mol/L NaOH | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...