Diisopropyl adipate ≥99.0% (by GC)
供应商: TCI
Synonyms:
Adipic acid diisopropyl ester
A0654-25ML
A0654-500ML
TCIAA0654-25MLEA
0
CNY
TCIAA0654-25ML
TCIAA0654-500ML
Diisopropyl adipate ≥99.0% (by GC)
Diisopropyl adipate
分子式:
C₁₂H₂₂O₄ 分子量: 230.30 g/mol 沸点: 120°C 溶点: –1°C 闪点: 124°C |
MDL编号:
MFCD00026391 CAS号: 6938-94-9 EINECS: 230-072-0 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...