N-Acetylneuraminic acid hydrate ≥98.0% (by titrimetric analysis)
供应商: TCI
Synonyms:
Neu5Ac, NANA, 5-Acetamido-3,5-dideoxy-D-glycero-D-galacto-nonulopyranosonic Acid
A0639-1G
A0639-100MG
A0639-5G
TCIAA0639-1GEA
0
CNY
TCIAA0639-1G
TCIAA0639-100MG
TCIAA0639-5G
N-Acetylneuraminic acid hydrate ≥98.0% (by titrimetric analysis)
N-Acetylneuraminic acid hydrate
分子式:
C₁₁H₁₉NO₉ 存储条件: Freezer |
MDL编号:
MFCD00006620 CAS号: 131-48-6 EINECS: 205-023-1 Merck Index: 14,08484 |
|
产品技术规格
Appearance | White to Orange to Green powder to crystal |
Purity (Neutralization titration) | min. 98.0% (calcd.on anh.substance) |
Specific rotation [a]20/D | -30.0 to -34.0 deg (C=1, H2O) |
Water | max. 12.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...