戊胺 mixture of isomers ≥98.0% (by GC)

供应商: TCI
Danger

Synonyms: 戊胺, n-戊胺, 1-氨基戊烷

A0446-25ML A0446-500ML
TCIAA0446-25MLEA 0 CNY
TCIAA0446-25ML TCIAA0446-500ML
戊胺 mixture of isomers ≥98.0% (by GC)
戊胺
分子式: H₃C(CH₂)₃CH₂NH₂
分子量: 87.16 g/mol
沸点: 104.4
溶点: -50
闪点: –1°C
MDL编号: MFCD00008236
CAS号: 110-58-7
UN号: 1106
ADR: 3 II
Merck Index: 13,00602
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79b7c3b68d8b3fbed377337bbc551c4b859f35c1&vid=748b1584dabec1f60b6f6a3713c868191d71fb75&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR