α-氨基-γ-丁内酯 ≥98.0% (by total nitrogen and titration analysis)

供应商: TCI
Warning

Synonyms: DL-高丝氨酸内酯, (±)-α-氨基-γ-丁内酯

A0283-1G A0283-5G
TCIAA0283-1GEA 0 CNY
TCIAA0283-1G TCIAA0283-5G
α-氨基-γ-丁内酯 ≥98.0% (by total nitrogen and titration analysis)
α-氨基-γ-丁内酯
分子式: C₄H₇NO₂
分子量: 182.02 g/mol
MDL编号: MFCD00012723
CAS号: 6305-38-0
EINECS: 228-614-6

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR