DL-allo-Threonine ≥98.0% (通过滴定分析) (contains Threonine)

供应商: TCI

A0214-1G A0214-100MG
TCIAA0214-1GEA 0 CNY
TCIAA0214-1G TCIAA0214-100MG
DL-allo-Threonine ≥98.0% (通过滴定分析) (contains Threonine)
DL-allo-Threonine
分子式: C₄H₉NO₃
MDL编号: MFCD00067248
CAS号: 144-98-9
EINECS: 205-645-3

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR