2,4,6-三溴-3-羟基苯甲酸 ≥97%, high purity
供应商: VWR Chemicals
Synonyms:
3-羟基-2,4,6-三溴苯甲酸
VWRC0817-100GEA
6363
CNY
VWRC0817-100G
VWRC0817-500G
2,4,6-三溴-3-羟基苯甲酸 ≥97%, high purity
2,4,6-三溴-3-羟基苯甲酸
分子式:
C₇H₃Br₃O₃ 分子量: 374.81 g/mol 沸点: 396 °C (1013 hPa) 溶点: 146 °C |
MDL编号:
MFCD00055557 CAS号: 14348-40-4 |
产品技术规格
Melting Range | 147°C - 149°C |
Purity | >=97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...