Barbituric acid, AnalaR NORMAPUR®

供应商: VWR Chemicals

VWRC86178.130EA 370 CNY
VWRC86178.130 VWRC86178.180
Barbituric acid, AnalaR NORMAPUR®
Barbituric acid
分子式: C₄H₄N₂O₃
MDL编号: MFCD00006666
CAS号: 67-52-7

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR